![Carnitine Acetyltransferase from pigeon breast muscle ammonium sulfate suspension, ≥50 units/mg protein](/deepweb/assets/sigmaaldrich/product/images/286/584/725d154a-e677-4bdc-ad0f-38487b38080c/640/725d154a-e677-4bdc-ad0f-38487b38080c.jpg)
Carnitine Acetyltransferase from pigeon breast muscle
![Carnitine Acetyltransferase from pigeon breast muscle ammonium sulfate suspension, ≥50 units/mg protein](/deepweb/assets/sigmaaldrich/product/images/286/584/725d154a-e677-4bdc-ad0f-38487b38080c/640/725d154a-e677-4bdc-ad0f-38487b38080c.jpg)
Synonym(s):
Acetyl-Co-A:carnitine-O-acetyltransferase, CrAT
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Tissue
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing