Skip to Content
MilliporeSigma
Carbinoxamine Maleate certified reference material, pharmaceutical secondary standard

Carbinoxamine maleate

Carbinoxamine Maleate certified reference material, pharmaceutical secondary standard
Synonym(s):
Carbinoxamine maleate salt
Empirical Formula (Hill Notation):
C16H19ClN2O · C4H4O4
CAS Number:
Molecular Weight:
406.86

Products

Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade