Skip to Content
MilliporeSigma
Carbenicillin Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered

Carbenicillin

Carbenicillin Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Synonym(s):
α-Carboxybenzylpenicillin solution
Empirical Formula (Hill Notation):
C17H18N2O6S
Molecular Weight:
378.40

Products

Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Mode of Action
Sterilization Method
Showing 1-1 of 1