Skip to Content
MilliporeSigma
CARAC Umicore, 98%

CARAC

CARAC Umicore, 98%
Synonym(s):
Dicarbonyl-acetylacetonato-rhodium(I), Dicarbonylrhodium(I) 2,4-pentanedionate, Rh(CO)2acac, Rhodium(I) dicarbonyl acetylacetonate
Linear Formula:
Rh(CO)2(C5H7O2)
CAS Number:
Molecular Weight:
258.03
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing