Skip to Content
MilliporeSigma
Camphorquinone 97%

Camphorquinone

Camphorquinone 97%
Synonym(s):
(±)-Camphorquinone, 2,3-Bornanedione
Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing