Skip to Content
MilliporeSigma
Calyculin A, Discodermia calyx InSolution, ≥98%, 100 µM in DMSO, Cell-permeable protein phosphatase 2A (PP2A) inhibitor

Calyculin A from Discodermia calyx

Calyculin A, Discodermia calyx InSolution, ≥98%, 100 µM in DMSO, Cell-permeable protein phosphatase 2A (PP2A) inhibitor
Synonym(s):
InSolution Calyculin A, Discodermia calyx
Empirical Formula (Hill Notation):
C50H81N4O15P
CAS Number:
Molecular Weight:
1009.17

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Research Area
Quality Segment