Skip to Content
MilliporeSigma
Calphostin C from Cladosporium cladosporioides ≥90% (HPLC), powder

Calphostin C from Cladosporium cladosporioides

Calphostin C from Cladosporium cladosporioides ≥90% (HPLC), powder
Synonym(s):
UCN-1028c
Empirical Formula (Hill Notation):
C44H38O14
CAS Number:
Molecular Weight:
790.76

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing