Skip to Content
MilliporeSigma
Callistephin chloride ≥95.0% (HPLC)

Callistephin chloride

Callistephin chloride ≥95.0% (HPLC)
Synonym(s):
3-(Glucosyloxy)-4′,5,7-trihydroxyflavylium chloride, Pelargonidin 3-O-glucoside chloride
Empirical Formula (Hill Notation):
C21H21ClO10
CAS Number:
Molecular Weight:
468.84

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing