Skip to Content
MilliporeSigma
Calix[4]arene-25,26,27,28-tetrol 95%

Calix[4]arene-25,26,27,28-tetrol

Calix[4]arene-25,26,27,28-tetrol 95%
Synonym(s):
Calix[4]arene
Empirical Formula (Hill Notation):
C28H24O4
CAS Number:
Molecular Weight:
424.49

Products

Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing