Skip to Content
MilliporeSigma
Calcium standard for AAS analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM

Calcium standard for AAS

Calcium standard for AAS analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Synonym(s):
Calcium chloride solution
Empirical Formula (Hill Notation):
CaCl2
CAS Number:
Molecular Weight:
110.98

Products

Brand
Formula Weight
Format
Element
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1