Skip to Content
MilliporeSigma
Calcium Ionophore A23187 ≥98% (TLC), powder

Calcium Ionophore A23187

Calcium Ionophore A23187 ≥98% (TLC), powder
Synonym(s):
Antibiotic A 23187, Calimycin
Empirical Formula (Hill Notation):
C29H37N3O6
CAS Number:
Molecular Weight:
523.62
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing