Skip to Content
MilliporeSigma
Calcium(II) bis(trifluoromethanesulfonimide) 95%

Calcium(II) bis(trifluoromethanesulfonimide)

Calcium(II) bis(trifluoromethanesulfonimide) 95%
Synonym(s):
Ca(NTf2)2, Calcium triflimide
Empirical Formula (Hill Notation):
C4CaF12N2O8S4
CAS Number:
Molecular Weight:
600.37

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing