Skip to Content
MilliporeSigma
Calcein Blue fluorescent dye

Calcein Blue

Calcein Blue fluorescent dye
Synonym(s):
4-Methylumbelliferone-8-methyliminodiacetic acid
Empirical Formula (Hill Notation):
C15H15NO7
CAS Number:
Molecular Weight:
321.28
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing