Skip to Content
MilliporeSigma
Cadmium perchlorate hydrate 99.999% trace metals basis

Cadmium perchlorate hydrate

Cadmium perchlorate hydrate 99.999% trace metals basis
Linear Formula:
Cd(ClO4)2·xH2O
CAS Number:
Molecular Weight:
311.31 (anhydrous basis)

Products

Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing