Skip to Content
MilliporeSigma
Cadmium nitrate tetrahydrate 99.997% trace metals basis

Cadmium nitrate tetrahydrate

Cadmium nitrate tetrahydrate 99.997% trace metals basis
Synonym(s):
Nitric acid, cadmium salt tetrahydrate
Linear Formula:
Cd(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
308.48
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing