Skip to Content
MilliporeSigma
Cadmium(II) acetate SAFC Hitech®, anhydrous, 99.995%

Cadmium(II) acetate

Cadmium(II) acetate SAFC Hitech®, anhydrous, 99.995%
Synonym(s):
Cadmium(II) acetate
Linear Formula:
Cd(OCOCH3)2
CAS Number:
Molecular Weight:
230.50

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Markush Class
Markush Group
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing