Skip to Content
MilliporeSigma
Butyltrimethylammonium bis(trifluoromethylsulfonyl)imide 99%

Butyltrimethylammonium bis(trifluoromethylsulfonyl)imide

Butyltrimethylammonium bis(trifluoromethylsulfonyl)imide 99%
Empirical Formula (Hill Notation):
C9H18F6N2O4S2
CAS Number:
Molecular Weight:
396.37

Products

Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing