Skip to Content
MilliporeSigma
Buprofezin PESTANAL®, analytical standard

Buprofezin

Buprofezin PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C16H23N3OS
CAS Number:
Molecular Weight:
305.44

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique