Skip to Content
MilliporeSigma
Bromopropylate PESTANAL®, analytical standard

Bromopropylate

Bromopropylate PESTANAL®, analytical standard
Synonym(s):
Isopropyl 4,4′-dibromobenzilate, Phenisobromolate
Empirical Formula (Hill Notation):
C17H16Br2O3
CAS Number:
Molecular Weight:
428.12
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique