Skip to Content
MilliporeSigma
(Bromomethyl)cyclohexane 99%

(Bromomethyl)cyclohexane

(Bromomethyl)cyclohexane 99%
Synonym(s):
Cyclohexylmethyl bromide
Linear Formula:
C6H11CH2Br
CAS Number:
Molecular Weight:
177.08

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing