![Bromodiphenhydramine hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/287/657/3564cff8-5a0b-40e6-820c-73f2b828efdf/640/3564cff8-5a0b-40e6-820c-73f2b828efdf.png)
Bromodiphenhydramine hydrochloride
![Bromodiphenhydramine hydrochloride United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/287/657/3564cff8-5a0b-40e6-820c-73f2b828efdf/640/3564cff8-5a0b-40e6-820c-73f2b828efdf.png)
Synonym(s):
2-[(RS)-(4-Bromophenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride, 2-[(4-Bromophenyl)phenylmethoxy]ethyl(dimethyl)ammonium chloride, Bromazine hydrochloride, Bromodiphenhydramine hydrochloride, NSC 36113
Empirical Formula (Hill Notation):
C17H20BrNO · HCl
CAS Number:
Molecular Weight:
370.71
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing