Skip to Content
MilliporeSigma
Bromocresol Green sodium salt crystalline

Bromocresol green sodium salt

Bromocresol Green sodium salt crystalline
Synonym(s):
Bromocresol green sodium salt, 3′,3″,5′,5″-Tetrabromo-m-cresolsulfonphthalein sodium salt
Empirical Formula (Hill Notation):
C21H13Br4NaO5S
CAS Number:
Molecular Weight:
720.00
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Agency
Chemical Composition
Quality Segment
Application
Special Grade