Skip to Content
MilliporeSigma
Bromobis(dimethylamino)borane 97%

Bromobis(dimethylamino)borane

Bromobis(dimethylamino)borane 97%
Synonym(s):
1-Bromo-N,N,N′,N′-tetramethylboranediamine
Linear Formula:
[(CH3)2N]2BBr
CAS Number:
Molecular Weight:
178.87

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing