Skip to Content
MilliporeSigma
BRIJ® O20 average Mn ~1,150

BRIJ® O20

BRIJ® O20 average Mn ~1,150
Synonym(s):
Polyoxyethylene (20) oleyl ether
Linear Formula:
C18H35(OCH2CH2)nOH, n~20
CAS Number:
Molecular Weight:
1149.53

Products

Brand
Formula Weight
Melting Point (°C)
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing