Skip to Content
MilliporeSigma
Brij® L23 solution 30 % (w/v) in H2O

Brij® L23 solution

Brij® L23 solution 30 % (w/v) in H2O
Synonym(s):
Brij® 35 solution, C12E23, Polyoxyethylene (23) lauryl ether
CAS Number:

Products

Product Category
Brand
Biological Source
Feature
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing