Skip to Content
MilliporeSigma
Borrelidin from Streptomyces parvulus, ≥98% (HPLC)

Borrelidin

Borrelidin from Streptomyces parvulus, ≥98% (HPLC)
Synonym(s):
2-(7-Cyano-8,16-dihydroxy-9,11,13,15-tetramethyl-18-oxooxacyclooctadeca-4,6-dien-2-yl)-cyclopentanecarboxylic acid, Borrelidine, Cyclopentanecarboxylic acid, NSC 216128, Treponemycin
Empirical Formula (Hill Notation):
C28H43NO6
CAS Number:
Molecular Weight:
489.64

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing