Skip to Content
MilliporeSigma
Boc-Trp-OH ≥99.0% (TLC)

Boc-Trp-OH

Boc-Trp-OH ≥99.0% (TLC)
Synonym(s):
Nα-(tert-Butoxycarbonyl)-L-tryptophan, Nα-Boc-L-tryptophan, Boc-Trp-OH
Empirical Formula (Hill Notation):
C16H20N2O4
CAS Number:
Molecular Weight:
304.34

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing