![Boc-Lys(Z)-OH (cryst) Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/118/488/f4276324-5e1a-4900-85f7-a3d67a1061da/640/f4276324-5e1a-4900-85f7-a3d67a1061da.jpg)
Boc-Lys(Z)-OH
![Boc-Lys(Z)-OH (cryst) Novabiochem®](/deepweb/assets/sigmaaldrich/product/images/118/488/f4276324-5e1a-4900-85f7-a3d67a1061da/640/f4276324-5e1a-4900-85f7-a3d67a1061da.jpg)
Synonym(s):
Boc-Lys(Z)-OH
Empirical Formula (Hill Notation):
C19H28N2O6
CAS Number:
Molecular Weight:
380.44
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing