Skip to Content
MilliporeSigma
Boc-Ile-OH ≥97%

Boc-Ile-OH

Boc-Ile-OH ≥97%
Synonym(s):
N-(tert-Butoxycarbonyl)-L-isoleucine, Boc-L-isoleucine
Linear Formula:
CH3CH2CH(CH3)CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
231.29
EC Number:

Products

Formula Weight
Melting Point (°C)
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing