Skip to Content
MilliporeSigma
Boc-Cys(Trt)-OH 97%

Boc-Cys(Trt)-OH

Boc-Cys(Trt)-OH 97%
Synonym(s):
N-(tert-Butoxycarbonyl)-S-trityl-L-cysteine, Nα-Boc-S-trityl-L-cysteine, Boc-Cys(Trt)-OH
Linear Formula:
(C6H5)3CSCH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
463.59

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing