![Boc-3-iodo-D-Ala-OMe 99%](/deepweb/assets/sigmaaldrich/product/structures/240/247/39e6b2eb-3c2e-4a85-9608-8b6fbf0fc534/640/39e6b2eb-3c2e-4a85-9608-8b6fbf0fc534.png)
Boc-3-iodo-D-Ala-OMe
![Boc-3-iodo-D-Ala-OMe 99%](/deepweb/assets/sigmaaldrich/product/structures/240/247/39e6b2eb-3c2e-4a85-9608-8b6fbf0fc534/640/39e6b2eb-3c2e-4a85-9608-8b6fbf0fc534.png)
Synonym(s):
N-(tert-Butoxycarbonyl)-3-iodo-D-alanine methyl ester, Boc-β-iodo-D-Ala-OMe, Boc-3-iodo-D-alanine methyl ester
Linear Formula:
ICH2CH[NHCO2C(CH3)3]CO2CH3
CAS Number:
Molecular Weight:
329.13
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing