Skip to Content
MilliporeSigma
Bis(triphenylphosphine)palladium(II) dichloride

Bis(triphenylphosphine)palladium(II) dichloride

Bis(triphenylphosphine)palladium(II) dichloride
Synonym(s):
Dichlorobis(triphenylphosphine)palladium(II), Palladium(II)bis(triphenylphosphine) dichloride, PdCl2(PPh3)2
Linear Formula:
[(C6H5)3P]2PdCl2
CAS Number:
Molecular Weight:
701.90
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Core