Skip to Content
MilliporeSigma
Bis(triphenylphosphine)nickel(II) dichloride synthesis grade

Bis(triphenylphosphine)nickel(II) dichloride

Bis(triphenylphosphine)nickel(II) dichloride synthesis grade
Synonym(s):
Dichlorobis(triphenylphosphine)nickel(II), Nickel(II)bis(triphenylphosphine) dichloride
Linear Formula:
[(C6H5)3P]2NiCl2
CAS Number:
Molecular Weight:
654.17
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing