Skip to Content
MilliporeSigma
Bis(trifluoromethane)sulfonimide lithium salt

Bis(trifluoromethane)sulfonimide lithium salt

Bis(trifluoromethane)sulfonimide lithium salt
Synonym(s):
Bis(trifluoromethylsulfonyl)amine lithium salt, Lithium bistrifluoromethanesulfonimidate
Linear Formula:
CF3SO2NLiSO2CF3
CAS Number:
Molecular Weight:
287.09
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Special Grade