Skip to Content
MilliporeSigma
Bis[tetrakis(hydroxymethyl)phosphonium] sulfate solution technical, 70-75% in H2O (T)

Bis[tetrakis(hydroxymethyl)phosphonium] sulfate solution

Bis[tetrakis(hydroxymethyl)phosphonium] sulfate solution technical, 70-75% in H2O (T)
Synonym(s):
Tetrakis(hydroxymethyl)phosphonium sulfate
Linear Formula:
[(HOCH2)4P]2(SO4)
CAS Number:
Molecular Weight:
406.28

Products

Formula Weight
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1