Skip to Content
MilliporeSigma
Bis(ethylenedithio)tetrathiafulvalene 98%

Bis(ethylenedithio)tetrathiafulvalene

Bis(ethylenedithio)tetrathiafulvalene 98%
Synonym(s):
BEDT-TTF, Bi[5,6-dihydro-1,3-dithiolo[4,5-b][1,4]-dithiine-2-ylidene]
Empirical Formula (Hill Notation):
C10H8S8
CAS Number:
Molecular Weight:
384.69

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing