
Bis(ethylcyclopentadienyl)ruthenium(II)

Synonym(s):
Diethylruthenocene
Linear Formula:
C7H9RuC7H9
CAS Number:
Molecular Weight:
287.36
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Reagent Type
Core
Showing 1-1 of 1