Skip to Content
MilliporeSigma
Bis(dicyclohexylphosphino)methane 95%

Bis(dicyclohexylphosphino)methane

Bis(dicyclohexylphosphino)methane 95%
Synonym(s):
Methylenebis(dicyclohexylphosphine)
Linear Formula:
(C6H11)2PCH2P(C6H11)2
CAS Number:
Molecular Weight:
408.58

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing