Skip to Content
MilliporeSigma
bisBenzimide H 33258 ≥98% (HPLC and TLC)

bisBenzimide H 33258

bisBenzimide H 33258 ≥98% (HPLC and TLC)
Synonym(s):
2′-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi(1H-benzimidazole) trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazoyl]-6-(1-methyl-4-piperazyl)benzimidazole trihydrochloride, BBIH, BXI-72, HOE 33258, Hoechst 33258, NSC334072
Empirical Formula (Hill Notation):
C25H24N6O · 3HCl
CAS Number:
Molecular Weight:
533.88
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Technique