![Bis(2,4,6-trichlorophenyl) oxalate BioReagent, suitable for chemiluminescence, ≥99.0% (AT)](/deepweb/assets/sigmaaldrich/product/structures/234/971/26bd6a13-ac95-4cad-8381-45f65638f776/640/26bd6a13-ac95-4cad-8381-45f65638f776.png)
Bis(2,4,6-trichlorophenyl) oxalate
![Bis(2,4,6-trichlorophenyl) oxalate BioReagent, suitable for chemiluminescence, ≥99.0% (AT)](/deepweb/assets/sigmaaldrich/product/structures/234/971/26bd6a13-ac95-4cad-8381-45f65638f776/640/26bd6a13-ac95-4cad-8381-45f65638f776.png)
Synonym(s):
2,4,6-Trichlorophenyl oxalate, Oxalic acid bis(2,4,6-trichlorophenyl) ester, TCPO
Empirical Formula (Hill Notation):
C14H4Cl6O4
CAS Number:
Molecular Weight:
448.90
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing