Skip to Content
MilliporeSigma
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%

Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate

Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate 98%
Empirical Formula (Hill Notation):
C17H24F3O3RhS
CAS Number:
Molecular Weight:
468.34

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing