Skip to Content
MilliporeSigma
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate
Empirical Formula (Hill Notation):
C16H24BF4Rh · xH2O
CAS Number:
Molecular Weight:
406.07 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core