Skip to Content
MilliporeSigma
Biphenyl-4-carbonyl chloride 97%

Biphenyl-4-carbonyl chloride

Biphenyl-4-carbonyl chloride 97%
Synonym(s):
4-Phenylbenzoyl chloride
Linear Formula:
C6H5C6H4COCl
CAS Number:
Molecular Weight:
216.66
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing