Skip to Content
MilliporeSigma
BICINE Redi-Dri™, ≥99%, free-flowing

BICINE

BICINE Redi-Dri™, ≥99%, free-flowing
Synonym(s):
N,N-Bis(2-hydroxyethyl)glycine
Empirical Formula (Hill Notation):
C6H13NO4
CAS Number:
Molecular Weight:
163.17

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Showing 1-3 of 3
Compare
Product Number
Description
Pricing