Skip to Content
MilliporeSigma
Bicalutamide (CDX) ≥98% (HPLC), powder

Bicalutamide (CDX)

Bicalutamide (CDX) ≥98% (HPLC), powder
Synonym(s):
Casodex, Cosudex, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide
Empirical Formula (Hill Notation):
C18H14F4N2O4S
CAS Number:
Molecular Weight:
430.37

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing