Skip to Content
MilliporeSigma
β-Estradiol 17-acetate ≥99%

β-Estradiol 17-acetate

β-Estradiol 17-acetate ≥99%
Synonym(s):
1,3,5(10)-Estratriene-3,17β-diol 17-acetate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 17-acetate
Empirical Formula (Hill Notation):
C20H26O3
CAS Number:
Molecular Weight:
314.42

Products

Product Category
Formula Weight
Physical Form
Purity
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing