Skip to Content
MilliporeSigma
BES sodium salt ≥99% (titration)

BES sodium salt

BES sodium salt ≥99% (titration)
Synonym(s):
N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid sodium salt
Empirical Formula (Hill Notation):
C6H14NNaO5S
CAS Number:
Molecular Weight:
235.23

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing