Skip to Content
MilliporeSigma
Benzyltrimethylammonium tribromide 97%

Benzyltrimethylammonium tribromide

Benzyltrimethylammonium tribromide 97%
Synonym(s):
BTMABr3, Benzyltrimethylammonium bromide dibromide
Linear Formula:
C6H5CH2N(Br3)(CH3)3
CAS Number:
Molecular Weight:
389.95

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing