Skip to Content
MilliporeSigma
Benzyloxyacetyl chloride 95%

Benzyloxyacetyl chloride

Benzyloxyacetyl chloride 95%
Linear Formula:
C6H5CH2OCH2COCl
CAS Number:
Molecular Weight:
184.62

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing