Skip to Content
MilliporeSigma
Benzyl ether ≥98%, FCC, FG

Benzyl ether

Benzyl ether ≥98%, FCC, FG
Synonym(s):
Dibenzyl ether
Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing